Order Entry
ContactUsLinkComponent
COVID-19 IgG/IgM Test, BTNX
COVID-19 IgG/IgM Test, BTNX
Catalog #: CA76469-800
Supplier:  BTNX INC.
CAS Number:  
Covid-19 Igg/Igm Rapid Antibody Test, Sample: Whole Blood / Serum / Plasma, Format: Cassette, Time to result: 15 minutes, Contents: Test Cassettes, Disposable Pipettes, Buffer, Product Insert, in vitro immunoassay for the direct and qualitative detection of anti-SARS-CoV-2 IgM/IgG, KT25
COVID-19 IgG/IgM Test, BTNX
Catalog #: CA76469-800
Supplier:  BTNX INC.
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    COVID-19 IgG/IgM test
  • Cat. no.:
    CA76469-800
  • Storage Temperature:
    2...30 °C (35.6...86 °F)

Specifications

About this item

The Rapid Response™ COVID-19 IgG/IgM Rapid Test Device is an in vitro immunoassay for the direct and qualitative detection of anti-SARS-CoV-2 IgM and anti-SARS-CoV-2 IgG in human whole blood (including venous whole blood and capillary whole blood), serum, or plasma.

  • IgG detection: Relative sensitivity 94.6% (CI: 84.2 to 98.6%), relative specificity: 100% (CI: 96.5 to 100%)
  • IgM detection: Relative sensitivity 94.6% (CI: 81.9 to 97.6%), relative specificity: 98.1% (CI: 93.3 to 99.5%)
  • Test time 15 minutes

At the Point of Care setting, this test is only authorized for use with fingerstick whole blood specimens. This test is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2, indicating recent or prior infection. The Rapid Response™ COVID-19 IgG/IgM Rapid Test Device should not be used for screening patients or to diagnose or exclude acute SARS-CoV-2 infection. At this time, it is unknown for how long antibodies persist following infection and if the presence of antibodies confers protective immunity. A negative or non-reactive result for an individual subject indicates absence of detectable COVID-19 virus antibodies. However, a negative or non-reactive rest result does not preclude the possibility of exposure to or infection with COVID-19 virus.

False positive results may occur due to cross-reactivity from pre-existing antibodies or other possible causes. Due to the risk of false positive results, confirmation of positive results should be confirmed using a second, different IgG/IgM detection method. Laboratories are required to report all results to the appropriate public health authorities. The test is for professional use only. This assay is not intended for home testing (or self-testing). The Rapid Response™ COVID-19 IgG/IgM Rapid Test Device may detect a response to vaccination.

Pursuant to section 5 of the Interim Order Respecting the Importation and Sale of Medical Devices for Use in Relation to COVID-19, made by the Minister of Health on March 18, 2020, the Rapid Response™ COVID-19 IgG/IgM Rapid Test Device is now authorized for sale or importation in Canada.

Customers Who Bought This Also Bought