Buchner Funnels, Table Top, PE, Perforated, 10.25", Chemglass
Supplier: Chemglass
Certificates
About this item
Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate.
- Welded hose barb connector accepts 12.7 mm (¹/₂") Int.Ø tube and threads into 3/4" NPT bung on the funnel
- Welded hose barbs allow the port to be as low as possible to the inside bottom of the funnel
- Fixed or removable perforated filter plates are made of 4.8 mm (³/₁₆") thick HDPE with 4.8 mm (³/₁₆") perforations on 11 mm (⁷/₁₆") centers
- Use temperature range maximum 52 °C (125 °F), minimum –20 °C (–4 °F)
Perforated filter plates available in two versions (fixed or removable); fixed for coarse filtration or use with a cloth or paper filter; removable for use in precious metals recovery.
Customers Who Bought This Also Bought