Buchner Funnels, Table Top, PE, Porous, 10.25", Chemglass
Supplier: Chemglass
Certificates
About this item
Filter large volumes quickly with a polyethylene table-top buchner funnel using your choice of filter plate.
- Welded hose barb connector accepts 12.7 mm (¹/₂") Int.Ø tube and threads into 3/4" NPT bung on the funnel
- Welded hose barbs allow the port to be as low as possible to the inside bottom of the funnel
- Fixed or removable filter plates are supported by a multiple ring grid
- HDPE, 6.4 mm (¹/₄") thick, with a non-porous ring at the periphery of the plate which seals filter paper when used with porous filter plates
- Use temperature range maximum 52 °C (125 °F), minimum –20 °C (–4 °F)
Available in two porosities (medium and coarse); medium is 45 to 90 µm while the coarse is 90 to 130 microns.
Customers Who Bought This Also Bought