Speci-Freez® Vaccine Thermo-Shippers, Com-Pac
Supplier: Com-Pac
Certificates
About this item
Specifically designed for shipping 9.02×9.02×1.6" vial trays (5 tray or 12 tray formats) which must maintain ultra-low temperature.
- Maintain –70 °C temperatures up to 48, 72, or 96 hours
- Custom sizes, capacities, and temperature performances available
- Use either dry ice pellets or blocks
Each shipper includes polystyrene cooler, insulative cooler liner, and 200 lbs. test outer carton.
Customers Who Bought This Also Bought