SP Wilmad-LabGlass MicroProbe/MicroCryoProbe MicroProbe/MicroCryoProbe NMR Tubes, SP Industries
Supplier: SP Industries
Certificates
About this item
These small volume NMR tubes meet or exceed the instrument manufacturers specifications.
- Tightest concentricity and camber in the industry
- Increased shimming quality and SNR
- Upper section Ext.Ø. is 5 mm
Wilmad has been manufacturing small volume NMR Tubes with the highest quality in the industry to meet the demand in small volume NMR. The Ultra-High Field MicroProbe Tube (>600 MHz) is 10 times more precise in terms of camber and concentricity than instrument manufacturers' stock tubes.
Customers Who Bought This Also Bought