VWR® SMS Coveralls, Made in USA
Supplier: VWR International
Certificates
About this item
VWR® Made in USA SMS Coveralls are an ideal option for single-use needs.
- 3-layer SMS construction
- Serged seams
- Produced with latex-free elastic
- Made in USA
Coveralls are manufactured from a unique multi-layered spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfortable, and breathable.
They provide light chemical splash resistance. The material is low-linting and resistant to tears and punctures. Ideally suited for use in high-class protection areas of medical, industrial, laboratory, electronic, and pharmaceutical environments.
Ordering information: Also available in the Made in USA PPE line are masks, lab coats, scrubs, and SMS coveralls.
Packaging: Packed 25 coveralls per case.
Specifications
- Catalog No:
- 76470-572
- 76470-568
- 76470-570
- 76470-550
- 76470-552
- 76470-546
- 76470-548
- 76470-558
- 76470-560
- 76470-554
- 76470-556
- 76470-562
- 76470-564
- 76457-138
- 76457-142
- 76457-140
- 76470-566
- 76470-528
- 76470-530
- 76470-526
- 76470-536
- 76470-538
- 76470-532
- 76470-534
- 76456-930
- 76470-544
- 76456-928
- 76470-540
- 76470-542
- 76456-936
- 76456-934
- 76456-940
- 76456-938
- 76456-944
- 76456-942
- 76456-948
- 76456-946
- 76456-932
- 76470-506
- 76470-508
- 76470-514
- 76470-516
- 76470-510
- 76470-512
- 76470-522
- 76470-524
- 76456-950
- 76470-518
- 76457-150
- 76470-520
- 76457-152
- 76456-958
- 76456-956
- 76456-962
- 76456-960
- 76456-966
- 76456-964
- 76456-970
- 76456-968
- 76456-954
- 76456-952
- 76470-494
- 76470-500
- 76470-502
- 76470-496
- 76470-498
- 76470-504
- 76456-900
- 76456-904
- 76456-902
- 76456-990
- 76456-988
- 76456-908
- 76456-974
- 76456-972
- 76456-906
- 76456-978
- 76456-976
- 76456-910
- 76456-982
- 76456-980
- 76456-986
- 76456-984
- 76456-914
- 76456-912
- 76456-918
- 76456-916
- 76456-922
- 76456-920
- 76456-926
- 76456-924
- 76457-146
- 76456-992
- 76457-144
- 76456-996
- 76456-994
- 76457-148
- 76457-000
- 76456-998
- 76457-002
- Garment application:
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Industrial, Electronics/semi-conductor, Laboratory, Pharmaceutical
Specifications
Related Information
Certificates
Search by Lot NumberTDS VWR® SMS Coveralls USA
Datasheet
VWR PPE Line VWR Protection Apparel
Product Literature
Customers Who Bought This Also Bought