Avantor® ACE® C18 UltraCore BIO, Analytical HPLC/UHPLC Columns
Supplier: Hichrom
Certificates
About this item
Avantor® ACE® UltraCore BIO C18 are high performing solid-core (core-shell) particle columns designed for a wide range of biomolecule applications. With both 300 and 500 Å pore sizes available, optimum pore size can be chosen. This phase is well suited to the analysis of more hydrophilic proteins over 5 kDa molecular weight.
- Wide pore, solid-core combination for excellent peak shape and resolution of large molecules
- High purity, base deactivated silica with excellent reproducibility
- Good choice for fast analysis of polypeptides
These stainless steel columns are available in a variety of dimensions and are highly compatible with LC-MS and high sensitivity applications.
Specifications
- Catalog No:
- 76457-864
- 76457-894
- 76457-852
- 76457-984
- 76457-846
- 76457-978
- 76457-936
- 76457-858
- 76457-942
- 76457-900
- 76457-954
- 76457-948
- 76457-906
- 76457-960
- 76457-918
- 76457-912
- 76457-870
- 76457-882
- 76457-876
- 76457-966
- 76457-930
- 76457-888
- 76457-972
- Carbon load:
- 1.4%
- 1.4%
- 1.4%
- 1.0%
- 1.4%
- 1.0%
- 1.0%
- 1.4%
- 1.0%
- 1.4%
- 1.0%
- 1.0%
- 1.4%
- 1.0%
- 1.0%
- 1.4%
- 1.4%
- 1.4%
- 1.4%
- 1.0%
- 1.0%
- 1.4%
- 1.0%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- 600 bar (8702.26 psi)
- pH range:
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- Surface area:
- 23 m²/g
- 23 m²/g
- 23 m²/g
- 16 m²/g
- 23 m²/g
- 16 m²/g
- 16 m²/g
- 23 m²/g
- 16 m²/g
- 23 m²/g
- 16 m²/g
- 16 m²/g
- 23 m²/g
- 16 m²/g
- 16 m²/g
- 23 m²/g
- 23 m²/g
- 23 m²/g
- 23 m²/g
- 16 m²/g
- 16 m²/g
- 23 m²/g
- 16 m²/g
- Temperature limit:
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
Specifications
Customers Who Bought This Also Bought