About this item
Tubes made of non-toxic polypropylene, caps made of polyethylene. Designed for storage and transportation of biological material.
- Good chemical resistance
- Good temperature resistance
- All graduated tubes are in 5 ml increments
Designed for storage and transportation of biological material. Tubes provide strength and clarity and exhibit some unique design features. Five sizes are available from 5 to 30 ml. They have external threads to provide a smooth and uniform inner surface.
A perfect leakproof seal is obtained by the use of a specially designed flexible sealing lip inside the polyethylene closures. Caps also feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn.