Specifications
- Description:Rapid ID test kit
- Cat. no.:76423-346
Specifications
About this item
ImmuView S. Pneumoniae and L. Pneumophila urinary antigen test kit, able to identify both S. Pneumoniae and Legionnaires disease at the same time-with just one test.
- Quick
- Accurate
- Easy diagnosis
Quick, accurate and easy diagnosis is vital because S. pneumoniae is the leading cause of community acquired pneumonia. L. pneumophila causes costly hospital and ICU admissions. Not only that, the two diseases are difficult to tell apart, a significant challenge, as treatments for each disease are different.