About this item
A40 Reflex® Coveralls are made of a microporous film and laminate fabric which provides a better liquid barrier than the leading competitor.
- Coveralls feature a respirator fit hood, zipper front, and elastic wrists and ankles
- Breathable, microporous film laminated non-woven fabric provides barrier protection against solid particles, dust, and liquid splash
- Reflex® design delivers enhanced comfort with fewer rip-outs
- 3.8X better liquid barrier than leading competitor
- 30% stronger fabric with up to 8X less lint than leading competitor
KleenGuard™ Personal Protective Equipment (PPE) enables rather than inhibits, allowing for maximum productivity and safety for staff and employees. KleenGuard™ A40 Reflex® coveralls with hood are a great addition when offering comfortable protective clothing is a must. Each garment is equipped with a respirator fit hood, zipper front, and elastic wrists and ankles. Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates, the KleenGuard™ A40 material provides a better liquid barrier and a better particulate barrier than leading competitors.
These coveralls with hood pass NFPA 99 for antistatic materials, pass ASTM F1670 testing for penetration of blood and bodily fluids and have Reflex® sizing for a more comfortable fit and less rip outs. Popular industries using this product include aviation, pharmaceutical manufacturing, liquid handling, general manufacturing, and much more.
Specifications
- Catalog No:
- CA76449-924
- CA76449-922
- CA76449-926
- CA76449-920
- CA76449-918
- Model:
- A40
- A40
- A40
- A40
- A40
- Anti-static:
- Yes
- Yes
- Yes
- Yes
- Yes
- Garment application:
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
Specifications
Related Information
TDS A40 Reflex® Coveralls
Datasheet
VWR® Focus Production Volume 3