ContactUsLinkComponent
Monocular Cordless LED Microscope, National
Monocular Cordless LED Microscope, National
Catalog #: 470329-172
Supplier:  National Optical
Microscope
Monocular Cordless LED Microscope, National
Catalog #: 470329-172
Supplier:  National Optical
Specifications
  • Description:
    Compound monocular microscope
  • Cat. no.:
    470329-172
  • Illumination:
    LED Cordless
Specifications
About this item
Quality at an affordable price.
  • DIN 4x, 10x, and 40xR, objectives are achromatic, parfocalled, parcentered, and color-coded
  • 45° inclined viewing head rotates 360° for easy repositioning
  • Single 8 hours charge allows up to 50 hours of use
The microscope that sets the standard for high school instruction, now with the freedom of rechargeable batteries. Includes all the most commonly specified features, like rugged metal construction and locked-on parts. Superior DIN optical system and precision mechanical components assure parfocalled and parcentered optical alignment for sharp, unsurpassed viewing. Utilizes rechargeable batteries for bright, cool illumination from long-life LED bulbs.
Customers Who Bought This Also Bought