VersaLoupe™ LH-30 Magnifier, Carson
Catalog #: 470312-386
Supplier: CARSON OPTICAL
Specifications
- Description:Magnifier, 10X
- Cat. no.:470312-386
Specifications
About this item
Accurate and easy to focus.
The LH-30 VersaLoupe™ is a 10x dual lens stand magnifier. It is a crystal clear acrylic loupe with an adjustable focus. Simply twist the eyepiece to focus. It is perfect for coins, stamps, and photography.
Customers Who Bought This Also Bought