ContactUsLinkComponent
VWR® Cleanroom Mask with Earloops, Made in USA
VWR® Cleanroom Mask with Earloops, Made in USA
Catalog #: 76451-956
Supplier:  VWR International
VWR* Clean Room Face Mask ( Non-Medical Use), Ear Loop, 99.79% Filtration Efficientcy, Metal Wire Nose Bridge
VWR® Cleanroom Mask with Earloops, Made in USA
Catalog #: 76451-956
Supplier:  VWR International
Specifications
  • Sterility:
    Non sterile
  • Color:
    White
  • Description:
    VWR® cleanroom mask with earloops
  • Cat. no.:
    76451-956
Specifications
About this item
The combination of strong BFE and PFE make these masks ideal for use in production or research areas.
  • Protection against bacteria and particles
  • 3-ply construction
  • Non sterile
  • With earloops
Part of the VWR® Made in USA PPE line, these cleanroom masks with earloops are a core item of any solid garment program, offering the adjustability user's desire.
Country of origin: USA.
Ordering information: Made in USA Masks with ties are also available; see catalog number 76409-318.
Packaging: Masks are packaged 50 per bag, 10 bags per case.
Customers Who Bought This Also Bought