HALO® Guard Column Holder, Advanced Materials Technology
Catalog #: 76445-284
Supplier: Advanced Materials Technology
Specifications
- Description:HALO® guard column holder, stainless steel
- Cat. no.:76445-284
Specifications
About this item
Guard columm holder for HALO® guard column cartridges (required).
Customers Who Bought This Also Bought