HALO® ES-CN, BIOCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
Certificates
About this item
HALO® BIOCLASS ES-CN Peptide columns made with innovative Fused-Core® particle technology are ideal for both ultrafast- and ultrahigh-resolution separations of peptides and polypeptides up to 20 kDa. HALO® peptide columns offer comparable peak capacity to sub-2 μm non-core columns at 40 to 50% back pressure (2.7 μm). HALO® 160 Å peptide columns are optimized for high performance in HPLC, UHPLC and LCMS peptide applications.
- Ideal for tryptic digests, post-translational modifications (PTMs), polypeptides
- HALO® Peptide ES-CN offers improved retention for polar peptides
- HALO® Peptide columns can be used in series to increase peak capacity for UHPLC and HPLC analyses of complex tryptic digest samples
- Particle Sizes: 2.7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
Alternate selectivities with HALO® Peptide ES-C18, ES-CN, and Phenyl-Hexyl phases.
HALO® guard columns are available for this phase.
Specifications
- Catalog No:
- 76444-250
- 76444-246
- 76443-548
- 76444-286
- 76445-328
- 76444-294
- 76444-290
- 76443-702
- 76443-698
- 76444-278
- 76444-274
- 76444-282
- 76444-270
- 76444-266
- 76444-258
- 76445-296
- 76444-254
- 76445-300
- 76445-288
- 76444-262
- 76444-330
- 76445-366
- 76446-752
- 76445-370
- 76444-326
- 76444-338
- 76446-744
- 76444-226
- 76444-334
- 76446-748
- 76446-750
- 76444-322
- 76445-362
- 76444-318
- 76445-354
- 76444-218
- 76445-358
- 76445-344
- 76444-306
- 76443-370
- 76445-348
- 76444-314
- 76443-378
- 76443-374
- 76444-310
- 76443-718
- 76444-302
- 76445-340
- 76443-706
- 76444-298
- 76445-332
- 76443-714
- 76443-710
- 76445-336
- 76443-544
- 76443-540
- 76445-396
- 76445-400
- 76445-388
- 76445-392
- 76445-384
- 76444-230
- 76445-374
- 76445-380
- 76444-238
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- Temperature limit:
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- USP code:
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
Specifications
Customers Who Bought This Also Bought