HALO® Diphenyl, BIOCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
Certificates
About this item
HALO® BIOCLASS Diphenyl 1000 Å protein columns made with innovative large pore Fused-Core® particle technology provide best-in-class performance for high-resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns.
- Ideal for mAbs, ADCs and large proteins due to unrestricted access to the bonded phase
- Best-in-class resolution for full characterization of mAbs and their variants
- Exceptional recovery performance and temperature stability from 40 to 90 °C
- Outstanding low bleed performance
HALO 1000 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
Alternate selectivities with HALO® Protein C4, Diphenyl and ES-C18 phases.
HALO® guard columns are available for this phase.
Specifications
- Catalog No:
- 76443-394
- 76444-418
- 76444-490
- 76444-412
- 76443-388
- 76443-400
- 76444-484
- 76444-406
- 76446-072
- 76446-078
- 76446-066
- 76443-382
- 76444-472
- 76444-478
- 76444-394
- 76444-466
- 76443-380
- 76444-400
- 76444-388
- 76446-060
- 76446-054
- 76444-460
- 76446-048
- 76444-454
- 76444-448
- 76444-382
- 76444-376
- 76537-468
- 76537-464
- 76537-460
- 76444-364
- 76444-508
- 76444-436
- 76444-442
- 76444-424
- 76444-344
- 76444-430
- 76444-502
- 76444-496
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Temperature limit:
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- USP code:
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11