HALO® Phenyl-Hexyl, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
Certificates
About this item
HALO® Phenyl-Hexyl columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å Phenyl-Hexyl columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
- Ideal for separating aromatic compounds using pi-pi interactions
- Alternate selectivity to alkyl phases
- Also available with 160 Å pores for peptide mapping
- Particle Sizes: 2, 2.7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® guard columns are available for this phase.
Specifications
- Catalog No:
- 76444-970
- 76443-824
- 76444-734
- 76443-884
- 76446-242
- 76444-714
- 76445-992
- 76445-752
- 76443-578
- 76444-950
- 76445-492
- 76446-222
- 76444-694
- 76445-972
- 76443-558
- 76443-864
- 76445-732
- 76445-812
- 76443-344
- 76445-008
- 76443-924
- 76445-872
- 76445-792
- 76443-324
- 76446-032
- 76444-990
- 76445-852
- 76443-598
- 76443-904
- 76446-012
- 76445-772
- 76445-048
- 76444-822
- 76443-984
- 76443-498
- 76446-162
- 76443-488
- 76443-452
- 76445-912
- 76443-484
- 76445-672
- 76443-448
- 76444-802
- 76444-044
- 76443-456
- 76445-832
- 76445-028
- 76443-964
- 76443-480
- 76444-862
- 76446-142
- 76443-466
- 76445-892
- 76443-944
- 76445-652
- 76445-412
- 76444-930
- 76443-434
- 76444-024
- 76443-424
- 76445-712
- 76445-472
- 76446-202
- 76444-674
- 76443-844
- 76445-952
- 76444-084
- 76443-416
- 76445-632
- 76444-842
- 76443-412
- 76444-602
- 76443-420
- 76444-004
- 76443-404
- 76446-182
- 76445-932
- 76445-692
- 76445-452
- 76444-064
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Temperature limit:
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- USP code:
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11