Resin Pellets Solid Matrix, Environmental Express®
Catalog #: 76437-108
Supplier: Environmental Express
CAS Number:
Specifications
- Description:Resin pellets solid matrix
- Cat. no.:76437-108
Specifications
About this item
Solid resin pellets for blanking and contamination-free confirmation.
- Demonstrate freedom from contamination
Contaminant-free resin pellets provide a clean, low-cost solid matrix blank.
Customers Who Bought This Also Bought