SimpleDist® Filter Membranes, Environmental Express®
Catalog # 76436-868
Supplier: Environmental Express
Specifications
- Description:SimpleDist® micro replacement top membranes
- Material:Plastic
- Environmentally Preferable:
- Cat. no.:76436-868
Specifications
About this item
Prevent sample-to-sample contamination.
- Resistant to strong acids and bases
Ideal replacement membrane filters for your radiological extration filter funnels and SimpleDist® micro tubes.