FlipMate™ Filtration Systems, Environmental Express®
Supplier: Environmental Express
Certificates
About this item
The FlipMate™ system easily filters the sample with the use of two threaded digestion cups, a filter assembly, vacuum tubing, and vacuum pressure source.
- Convenient, economical method for filtration
- Produce quality results with clean samples
- Results within seconds
The sample is placed in one cup, the filter assembly threaded onto the top of the cup, then the cup and filter are flipped, the receiving cup attached, vaccum tubing attached, vaccum pressure applied, and the sample is then pulled from the sample cup, through the filter assembly, and into the empty cup.
Delivery: Each filter is supplied in an aluminum dish that has the weight and filter ID in print and barcode formats.
Customers Who Bought This Also Bought