4-Aminobutyric acid ≥99%, Biotech Reagent
Catalog #: CAJTV265-10
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 56-12-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:10 kg
Specifications
Specification Test Results
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Appearance (White to off-white solid):Passes Test
- Identification (FTIR):Passes Test
- Clarity of solution (5% in water):Passes Test
- Assay:≥99 %
- Loss on drying (105°C):≤0.5 %
- Residual solvent - Methanol:≤3000 ppm
- Residual solvent - Ethanol:≤5000 ppm
- Residual solvent - Isopropanol:≤5000 ppm
- Aluminum (Al):≤60 ppm
- Cadmium (Cd):≤2 ppm
- Chromium (Cr):≤7 ppm
- Cobalt (Co):≤100 ppm
- Copper (Cu):≤20 ppm
- Manganese (Mn):≤10 ppm
- Zinc (Zn):≤900 ppm
- Nickel (Ni):≤20 ppm
- Selenium (Se):≤25 ppm
- Bacterial endotoxins, EU/g (For information only):
- Total Aerobic Microbial Count, For Information Only:
- Total Yeasts and Molds Count, For Information Only:
- Residual Solvents - Only Class 3 solvents are likely to be present from:
- the product purification process. Class 3 solvents are identified,:
- quantified and reported for each production batch.:
- 4-Aminobutyric acid ≥99%, Biotech Reagent:
Specification Test Results
About this item
Synonyms: GABA
Melting Pt:195…204 °C
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 195…204 °C
:Flash Pt:
:Flash Pt:
:M.W. 103.12 g/mol
:4-Aminobutyric acid
:248
:M.O.C. Category R
Formula:C₄H₉NO₂
MW:103.12 g/mol
MDL Number:MFCD00008226
CAS Number:56-12-2
Customers Who Bought This Also Bought