pZerve DMSO Free Cryopreservation Solutions, Protide Pharmaceuticals
Supplier: PROTIDE PHARMACEUTICALS
Certificates
About this item
A unique cryopreservation solution that does not contain DMSO, fetal bovine serum, or animal protein. pZerve™ is ready to use and does not require any dilution or further processing.
- Does not contain DMSO
- No fetal bovine serum or other animal proteins
- Sterile/Ready to use
Certifications: cGMP manufactured.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.