To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Linearity FLQ TDM for Roche
- Size:5×4 ml
- Cat. no.:76446-934
- Stability:5 days when stored at 2...8 °C
- Analyte:Acetominophen, Amikacin, Carbamezepine. Digoxin, Gentamicin, NAPA, Phenobarbital, Phenytoin, Proccainamide, Quinidine, Salicylate, Theophylline, Tobramycin, Valproic Acid, Vancomycin
- Type:Frozen liquid
Specifications
About this item
Linearity FLQ TDM for Roche systems is an IVD device consisting of five levels.
- Format: Frozen liquid
It is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: acetominophen, amikacin, carbamezepine. digoxin, gentamicin, NAPA, phenobarbital, phenytoin, proccainamide, quinidine, salicylate, theophylline, tobramycin, valproic acid, vancomycin.