Fluorescent Viewing Goggles, Noir medicals
Catalog #: 76420-054
Supplier: NOIR MEDICAL
Specifications
- Description:Fluorescent viewing goggles
- Cat. no.:76420-054
Specifications
About this item
Necessary protection when operating forensic light source.
Completely shield your eyes from UV A and UV B rays with these flexible orange viewing goggles. Clearly see latent prints and other evidence fluorescing with our wrap around goggles that meet ANSI standards and comfortably fit over prescription glasses.
Customers Who Bought This Also Bought