Order Entry
ContactUsLinkComponent
C11 through C18 Alkanes Mix, SPEX CertiPrep
C11 through C18 Alkanes Mix, SPEX CertiPrep
Catalog #: SXC11-18
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
C11 through C18 Alkanes Mix, SPEX CertiPrep
Catalog #: SXC11-18
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    Methylene chloride
  • Packaging:
    Ampoule, amber
  • Description:
    C11 through C18 Alkanes Mix
  • Size:
    1 ml

Specifications

About this item

Petroleum and LUST Standards

The EPA regulates the monitoring and testing of underground storage tanks (USTs), that were or are used to store petroleum or other hazardous substances, which could impact the environment and human health. Currently, there are over half a million registered underground storage tanks in the US alone. Many tanks installed prior to the mid-80s were composed of steel, which can corrode over time allowing for leakage into the surrounding groundwater and soil.

The Hazardous and Solid Waste Amendments to the Resource Conservation and Recovery Act (RCRA), passed in 1984, required the US Environmental Protection Agency (EPA) to develop regulations for the underground storage of motor fuels. The EPA requires owners and operators of USTs to maintain and clean up sites damaged by petroleum contamination.

SPEX CertiPrep provides standards designed to meet the testing requirements set up by the EPA and many individual state regulations for LUST and LUFT.

Customers Who Bought This Also Bought