To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:1000 µg/ml
- Matrix:Hexane
- Packaging:Ampoule, amber
- CAS No.:11097-69-1
- Description:Aroclor 1254
- Size:1 ml
- Description:Aroclor 1254
- Cat. no.:SXPCB-1254H
Specifications
About this item
Semivolatiles
US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.