To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:1000 µg/ml
- Matrix:Acetone
- Packaging:Ampoule, amber
- CAS No.:52645-53-1
- Description:Permethrin (cis & trans)
- Size:1 ml
- Description:Permethrin (cis & trans)
- Cat. no.:SXS-3000
Specifications
About this item
Pesticides, PCBs and Herbicides
The EPA is tasked with the monitoring of environmental systems as they pertain to contamination and human health. The methods issued by the EPA are created to respond to specific toxins or persistent organic pollutants (POPs) found in environmental samples such as soil, source water, drinking water, and waste. In particular, pollutants such as polychlorinated biphenyls (PCBs), which were in widespread industrial use up until their restriction, are of concern due to their stable and persistent nature in the environment. Another group of chemicals of high concern is the hundreds of commercial pesticides and herbicides in use in the world today. Pesticides, from algaecides to virucides, are used in large quantities in industrial and private agriculture. The concern over human pesticide exposure over the past few decades has led to extensive monitoring of these pesticides. It has been reported that over 98% of insecticides and 95% of herbicides affect areas other than their intended target product. It is essential that monitoring agencies have accurate standard mixes to measure the pesticide levels in the environment.
Many new pesticides are now being tested using highly sensitive LC/MS techniques, in addition to traditional GC techniques, to determine minute amounts of residue in environmental samples and food products.
At SPEX CertiPrep, we facilitate ease of monitoring and testing of pesticides by creating pesticide test mixes to suit your monitoring needs. SPEX CertiPrep is the leader in offering pesticide standards designed to work within EPA, AOAC and FDA analytical testing methods using all of the leading analytical techniques: LC, LC/MS, GC, and GC/MS. Many pesticide standard mixes are readily available in our catalog along with a large list of single pesticide standards. In addition, custom pesticide mixes can be made to your specifications to create a mix that meets your needs.
US EPA Method 8081 is an analytical method for the monitoring of organochlorine pesticides and PCBs in environmental samples by GC/ECD