Avantor® Hichrom HI-DEX DMP Beta, GC Columns
Supplier: VWR International
Certificates
About this item
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76429-990
- 76429-992
- 76429-986
- 76429-988
- 76429-982
- 76429-984
- 76429-978
- 76429-980
- 76429-976
- 76430-080
- Phase name:
- HI-DEX DMP Beta
- HI-DEX DMP Beta
- HI-DEX DMP Beta
- HI-DEX DMP Beta
- HI-DEX DMP Beta
- HI-DEX DMP Beta
- HI-DEX DMP Beta
- HI-DEX DMP Beta
- HI-DEX DMP Beta
- HI-DEX DMP Beta
- Phase composition:
- Dimethyl-pentyl Beta Cyclodexstrin based
- Dimethyl-pentyl Beta Cyclodexstrin based
- Dimethyl-pentyl Beta Cyclodexstrin based
- Dimethyl-pentyl Beta Cyclodexstrin based
- Dimethyl-pentyl Beta Cyclodexstrin based
- Dimethyl-pentyl Beta Cyclodexstrin based
- Dimethyl-pentyl Beta Cyclodexstrin based
- Dimethyl-pentyl Beta Cyclodexstrin based
- Dimethyl-pentyl Beta Cyclodexstrin based
- Dimethyl-pentyl Beta Cyclodexstrin based
- Polarity:
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Crossbond:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Temperature limit:
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C