Avantor® Hichrom HI-PLUS 75, GC Columns
Supplier: VWR International
Certificates
About this item
Offering a range of selectivities with differing copolymer rations of PEG and methyl polysiloxane. HI-PLUS, HI-PLUS 10, HI-PLUS 25, HI-PLUS 75, HI-PLUS 90.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76430-738
- 76427-760
- 76427-926
- 76427-922
- 76427-924
- 76430-740
- 76427-918
- 76427-920
- 76430-742
- 76427-914
- 76427-916
- 76427-910
- 76427-912
- 76427-908
- 76427-964
- 76430-716
- 76427-960
- 76430-718
- 76427-962
- 76430-734
- 76430-736
- 76430-730
- 76430-732
- 76430-726
- 76427-904
- 76430-728
- 76427-906
- 76430-722
- 76427-900
- 76427-902
- 76430-724
- 76427-896
- 76430-720
- 76427-898
- 76427-892
- 76427-894
- 76427-888
- 76427-890
- 76427-784
- 76427-786
- 76427-802
- 76427-804
- 76431-346
- 76427-798
- 76431-348
- 76427-800
- 76431-342
- 76427-794
- 76431-344
- 76427-796
- 76427-790
- 76427-792
- 76427-788
- 76427-766
- 76427-762
- 76427-764
- 76427-780
- 76427-782
- 76427-776
- 76427-778
- 76427-772
- 76427-774
- 76427-768
- 76427-770
- 76427-846
- 76427-842
- 76427-844
- 76427-838
- 76427-840
- 76427-834
- 76427-836
- 76427-830
- 76427-832
- 76427-828
- 76427-806
- 76427-824
- 76427-826
- 76427-820
- 76427-822
- 76427-816
- 76427-818
- 76427-812
- 76427-814
- 76427-808
- 76427-810
- 76427-946
- 76427-942
- 76427-944
- 76427-938
- 76427-940
- 76430-712
- 76427-956
- 76430-714
- 76427-958
- 76427-886
- 76427-952
- 76430-710
- 76427-954
- 76427-882
- 76427-948
- 76427-884
- 76427-950
- 76427-878
- 76427-880
- 76427-874
- 76427-876
- 76427-870
- 76427-872
- 76427-868
- 76427-934
- 76427-936
- 76427-864
- 76427-930
- 76427-866
- 76427-932
- 76427-860
- 76427-862
- 76427-928
- 76427-856
- 76427-858
- 76427-852
- 76427-854
- 76427-848
- 76427-850
- Phase name:
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- HI-PLUS 75
- Phase composition:
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Copolymer 75% Polyethylene glycol (PEG), 25% Methyl Polysiloxane
- Polarity:
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C