Avantor® Hichrom HI-DEX B-03, GC Columns
Supplier: VWR International
Certificates
About this item
Unique selectivity for the separation of enantiomers. Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76429-858
- 76430-056
- 76429-854
- 76429-856
- 76430-054
- 76429-872
- 76429-868
- 76429-870
- 76429-864
- 76429-866
- 76429-860
- 76429-862
- Phase name:
- HI-DEX B-03
- HI-DEX B-03
- HI-DEX B-03
- HI-DEX B-03
- HI-DEX B-03
- HI-DEX B-03
- HI-DEX B-03
- HI-DEX B-03
- HI-DEX B-03
- HI-DEX B-03
- HI-DEX B-03
- HI-DEX B-03
- Phase composition:
- Proprietary Cyclodexstrin based
- Proprietary Cyclodexstrin based
- Proprietary Cyclodexstrin based
- Proprietary Cyclodexstrin based
- Proprietary Cyclodexstrin based
- Proprietary Cyclodexstrin based
- Proprietary Cyclodexstrin based
- Proprietary Cyclodexstrin based
- Proprietary Cyclodexstrin based
- Proprietary Cyclodexstrin based
- Proprietary Cyclodexstrin based
- Proprietary Cyclodexstrin based
- Polarity:
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Crossbond:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Temperature limit:
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C