Avantor® Hichrom HI-PLUS 10, GC Columns
Supplier: VWR International
Certificates
About this item
Offering a range of selectivities with differing copolymer rations of PEG and methyl polysiloxane. HI-PLUS, HI-PLUS 10, HI-PLUS 25, HI-PLUS 75, HI-PLUS 90.
Maximum temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76427-404
- 76427-406
- 76427-400
- 76427-402
- 76427-396
- 76427-398
- 76427-392
- 76427-394
- 76427-388
- 76427-390
- 76427-408
- 76427-386
- 76427-382
- 76427-384
- 76427-378
- 76427-380
- 76427-374
- 76427-376
- 76427-370
- 76427-372
- 76427-368
- 76427-444
- 76427-446
- 76427-440
- 76427-442
- 76427-436
- 76427-438
- 76431-332
- 76427-432
- 76427-434
- 76427-450
- 76427-452
- 76427-448
- 76427-426
- 76427-422
- 76427-424
- 76427-418
- 76427-420
- 76427-414
- 76427-416
- 76427-410
- 76427-412
- 76431-328
- 76427-428
- 76431-330
- 76427-430
- 76431-326
- 76430-658
- 76427-484
- 76427-550
- 76427-486
- 76427-552
- 76430-654
- 76427-480
- 76427-548
- 76427-482
- 76430-656
- 76430-650
- 76427-476
- 76430-652
- 76427-478
- 76430-668
- 76427-494
- 76427-496
- 76430-670
- 76430-664
- 76427-490
- 76430-666
- 76427-492
- 76430-660
- 76430-662
- 76427-488
- 76427-544
- 76427-546
- 76427-540
- 76427-542
- 76427-536
- 76427-538
- 76427-466
- 76427-532
- 76427-534
- 76427-462
- 76427-528
- 76427-464
- 76427-530
- 76427-458
- 76427-460
- 76427-454
- 76427-456
- 76430-646
- 76427-472
- 76430-648
- 76427-474
- 76430-642
- 76427-468
- 76430-644
- 76427-470
- 76427-364
- 76427-366
- 76427-360
- 76427-362
- 76427-356
- 76427-358
- 76427-352
- 76427-354
- 76427-524
- 76427-348
- 76427-350
- 76427-526
- 76427-520
- 76427-522
- 76427-506
- 76427-502
- 76427-504
- 76430-672
- 76427-498
- 76430-674
- 76427-500
- 76427-516
- 76427-518
- 76427-512
- 76427-514
- 76427-508
- 76427-510
- Phase name:
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- HI-PLUS 10
- Phase composition:
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Copolymer 10% Polyethylene glycol (PEG), 90% Methyl Polysiloxane
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes