Avantor® Hichrom HI-PLUS 25, GC Columns
Supplier: VWR International
Certificates
About this item
Offering a range of selectivity's with differing copolymer rations of PEG and methyl polysiloxane. HI-PLUS, HI-PLUS 10, HI-PLUS 25, HI-PLUS 75, HI-PLUS 90.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76427-646
- 76427-642
- 76427-644
- 76427-638
- 76427-640
- 76427-744
- 76427-634
- 76427-746
- 76427-636
- 76427-630
- 76427-740
- 76427-632
- 76427-742
- 76427-648
- 76427-758
- 76427-650
- 76427-754
- 76427-756
- 76427-750
- 76427-752
- 76427-748
- 76427-624
- 76427-626
- 76427-620
- 76427-622
- 76427-616
- 76427-618
- 76427-612
- 76427-614
- 76427-608
- 76427-610
- 76427-736
- 76427-628
- 76427-738
- 76427-732
- 76427-734
- 76427-728
- 76427-730
- 76427-686
- 76427-682
- 76427-684
- 76427-678
- 76431-336
- 76431-338
- 76427-680
- 76427-674
- 76431-334
- 76427-676
- 76427-692
- 76427-694
- 76427-688
- 76427-690
- 76431-340
- 76427-664
- 76427-666
- 76427-660
- 76427-662
- 76427-656
- 76427-658
- 76427-652
- 76427-654
- 76427-670
- 76427-672
- 76427-668
- 76427-566
- 76427-562
- 76427-564
- 76427-558
- 76427-560
- 76427-554
- 76427-556
- 76427-726
- 76427-722
- 76427-724
- 76427-718
- 76427-720
- 76427-704
- 76427-706
- 76427-700
- 76427-702
- 76427-696
- 76427-698
- 76427-714
- 76427-716
- 76427-710
- 76427-712
- 76427-708
- 76427-606
- 76427-602
- 76427-604
- 76427-598
- 76427-600
- 76427-594
- 76427-596
- 76430-698
- 76427-590
- 76427-592
- 76430-694
- 76430-696
- 76427-588
- 76430-708
- 76430-704
- 76430-706
- 76430-700
- 76430-702
- 76427-584
- 76427-586
- 76427-580
- 76427-582
- 76427-576
- 76427-578
- 76427-572
- 76427-574
- 76430-676
- 76427-568
- 76430-678
- 76427-570
- 76430-690
- 76430-692
- 76430-686
- 76430-688
- 76430-682
- 76430-684
- 76430-680
- Phase name:
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- HI-PLUS 25
- Phase composition:
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Copolymer 25% Polyethylene glycol (PEG), 75% Methyl polysiloxane
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C