To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:1 µg/ml
- Element:Platinum
- Matrix:2% HCl
- Packaging:LDPE bottle
- Size:125 ml
Specifications
About this item
Single element standard for ICP-MS.
- Made with an acid and ASTM Type I Water
- Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
- Directly traceable to NIST (where applicable)
Claritas PPT® Grade CRMs are designed for ICP and ICP-MS analysis. They are available in single and multi-element solutions. The standards are set at 1 µg/ml, 100 µg/ml, or 1,000 µg/ml and packaged in 30 ml and 125 ml bottles to minimize contamination. They are made using ultra high purity acids, the highest grade starting materials and high purity water in order to minimize contaminants.
Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016