Acetonitrile ≥99.8%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: Methyl cyanide,Ethanoic acid nitrile,Cyanomethane
Formula:H₃CCN
MW:41.05 g/mol
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
MDL Number:MFCD00001878
CAS Number:75-05-8
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 81.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –45.7 °C
:Flash Pt:
:Flash Pt: 2 °C (closed cup)
:M.W. 41.05 g/mol
:Acetonitrile
EINECS:200-835-2
:81.6 °C (1013 hPa)
UN:1648
REACH:01-2119471307-38
:3
:II
:2 °C
:GHS02
:GHS07
Storage Temperature:Ambient
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (CH₃CN) (by GC, corrected for water):≥99.8 %
- Appearance (clear, colorless liquid):Passes Test
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤0.0002 %
- ACS - Titrable Acid (µeq/g):≤8
- ACS - Titrable Base (µeq/g):≤0.6
- ACS - Water (H₂O)(by Karl Fischer titrn):≤0.02 %
- ACS - HPLC Gradient Elution (No Peak Greater than 25% of Full Scale)(0.005 au):Passes Test
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 190 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 200 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:≤0.02
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 - 400 nm:≤0.005
- Filtered through a 0.2 micron filter.:
- Acetonitrile ≥99.8%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™:
Specification Test Results
Related Information
Certificates
Search by Lot Number2856 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Macron 2856.pdf
Datasheet
sds_MACR2856
SDS
Customers Who Bought This Also Bought