BioClean™ Cut Resistant Glove Liner, S-BCRL, Ansell
Supplier: Avantor
Certificates
About this item
BioClean™ sterile cut-resistant glove liners feature Dyneema® Diamond yarn and provide cut resistance and protection during rigorous procedures.
- Optimal dexterity, comfort, and fit
- Recommended to be worn between two cleanroom gloves
- Ultra high molecular weight polyethylene material
- Supplied sterile via gamma irradiation
- Latex-free, contains no NRL proteins
These cut-resistant glove liners are designed to be worn between two cleanroom gloves to provide EN 388 Level B and ANSI A2 cut protection when handling sharp objects or cleaning apparatus which pose a cut risk. Ideally suited for use in life science, chemical, and healthcare industries.
Glove liners can be autoclaved for up to 5 cycles at 121 °C for 30 minutes.
Certifications: Mechanical protection EN388:2016 Level B and ANSI A2. Glove Standard EN 420:2003+A1:2009.
Packaging: Glove liners are packaged one pair per inner PE wallet; one wallet per sealed EasyTear PE pouch; 10 pouches per sealed outer PE bag; 10 outer bags per lined carton.
Specifications
- Catalog No:
- 76326-070
- 76326-144
- Model:
- S-BCRL
- S-BCRL
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Finish:
- Knitted
- Knitted
- Sterility:
- Sterile
- Sterile