VWR® Finger Textured Nitrile Examination Gloves
Supplier: VWR International
Certificates
About this item
Ambidextrous gloves are extra-soft and comfortable for everyday use. They protect hands from a broad range of chemicals.
- Non sterile
- Extra-soft and comfortable
- Powder-free
- Beaded cuff allows easy donning
Ideal for use in industrial and laboratory applications. Made of premium compounded nitrile for high manufacturing quality measures and product consistency. Gloves protect against biohazards and a broad range of chemicals. Composition is 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins. Finger textured gloves feature rolled, beaded cuffs.
Certifications: Gloves meet or exceed ASTM D6319 on water leak and dimension. Manufactured under GMP, ISO 9001, ISO 13485, and MDSAP quality system.
Packaging: 100 gloves per box, 10 boxes per case.
Specifications
- Catalog No:
- 76411-504
- Ambidextrous/Hand-specific:
- Ambidextrous
- Latex/Latex-free:
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Material:
- Nitrile
- Finish:
- Textured
- Cuff style:
- Beaded
- Sterility:
- Non sterile