About this item
Complete print-on-demand solutions that can be customized to specific application requirements.
- Highly versatile cleanroom printer
- Handles most labeling and sign making tasks
- Easy to set up and ready to go right out of the box
- Fast, high resolution 600 dpi printing
The UltraLabel PRO-600 series cleanroom label printer is designed for critical environment applications. It offers enhanced functionality, including print resolution of 600 dpi and a user-friendly TFT display. The high-resolution label printer supports a range of label-making applications including 2D, barcoding, serialization, inventory control identification, product labels, pipe markers, and GHS labels. The UltraLabel PRO-600 is compact and versatile and can easily be integrated into any cleanroom environment. The printer and all related supplies are cleanroom certified and packaged to keep them free from contaminants.