Double-Sided Polyester Removable Adhesion Cleanroom Tape, UltraTape
Catalog #: 11212-423
Supplier: Ultratape Industries
Specifications
- Length:36 yards
- Width:0.75"
- Color:Clear
- Material:UltraClean
- Cat. no.:11212-423
- Supplier No.:1520CL075P3D
- Adhesive material:Acrylic
Specifications
About this item
Particle-free, residue-free tapes.
- Double-sided removable adhesive tape
Customers Who Bought This Also Bought