ContactUsLinkComponent
Cleanroom Barrier Tape, 'Danger', UltraTape
Cleanroom Barrier Tape, 'Danger', UltraTape
Catalog #: 89203-777
Barricade Tape
Cleanroom Barrier Tape, 'Danger', UltraTape
Catalog #: 89203-777
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    1000'
  • Width:
    3"
  • Color:
    Red with black print
  • Material:
    Classic clean
  • Cat. no.:
    89203-777
  • Supplier No.:
    0422RE300P1S
  • Adhesive material:
    Non-adhesive
Specifications
About this item
Particle-free, residue-free tapes.
  • Red and black barrier tape
  • Printed with 'DANGER DO NOT ENTER'
  • Barrier tapes are used to section off dangerous areas from general access
Non-adhesive caution barricade tapes are made with polyethylene film on plastic cores, and printed with easy-to-read letters.
Customers Who Bought This Also Bought