Flip Top Inlet Sealing System for GC,Agilent Technologies
Catalog #: CA76508-186
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Inlet liner O-ring, non-stick, for Agilent Flip Top Inlet
- Cat. no.:CA76508-186
Specifications
About this item
The flip-top inlet sealing system allows you to safely and reliably change an inlet liner, without tools, in a consistent leak-free manner.
- Installs easily in 15 minutes
- Cuts liner replacement time to as little as 30 seconds
- No more frustrating searches for special wrenches or tools
- No more handling of heated parts, no more burns or scrapes
- Minimizes exposure to ambient air, extending column life
Agilent’s flip-top inlet sealing system allows you to safely and reliably change an inlet liner in as little as 30 seconds, without tools, in a consistent leak-free manner. Available exclusively from Agilent, this device has a levered arm that attaches to any 6890/6850/5890 insert weldment and locks to the injection port using an adapter ring screwed onto the inlet. Once the device is installed, you simply lift the arm, which releases the insert weldment from the injection port, and allows instant access to the liner. Simply reverse the process to reseal the weldment to the port.
Packaging: Packaged for convenience and cleanliness in a novel dial package that delivers one clean O-ring at a time.
Customers Who Bought This Also Bought