ASX-500 Rinse Reservoirs and Other Autosampler Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Rinse reservoirs and other autosampler supplies used with Agilent SPS 4 and ASX 500 autosamplers.
- Upgrade probe rinsing capacity, giving you flexibility to improve the capabilities of the autosamplers used in your lab
- Convert your probe rinsing option to a pumped drain to allow liquid overflow to be pumped from the rinse station for more through rinsing of the probe
- Increase rinsing capacity with a high volume rinse kit, to ensure compatibility with switching valves such as the Agilent AVS 6/7, SVS 2+, or ISIS 3
- Change the components in the rinse path to organic resistant materials, to enable operation with organic solvents
A number of different autosamplers have been used with Agilent AA, ICP-OES, MP-AES, and ICP-MS instruments, depending on the specific model. This includes the Agilent SPS 4 used with current instruments, and the ASX 500 Series used with earlier model instruments. Each model autosampler is designed to automate the presentation of liquid samples, drawn from tubes held in racks loaded on the rack location mat, to the instrument for analysis.;A range of rinse reservoirs and other autosampler supplies are available to upgrade probe rinsing capacity, giving you flexibility to improve the capabilities of the autosamplers used in your lab.
Customers Who Bought This Also Bought