CrossLab Pump Supplies for Thermo/Dionex LC Systems, Agilent Technologies
Catalog #: CAAG8002-0516
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Agilent check valve cartridge, sapphire, biocompatible, for Dionex UltiMate pumps
- Cat. no.:CAAG8002-0516
Specifications
About this item
Pump supplies for selected Thermo Fisher Scientific LC systems.
- Agilent CrossLab parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent
Agilent pump supplies for Thermo Fisher Scientific HPLC systems include the most important components of a pump: seals and check valves.
Regular pump maintenance helps lower operating costs and ensure precise and consistent results. By following a regular maintenance routine, you can count on maximum uptime, steady and accurate solvent flow, and pressure stability throughout the lifetime of the pump.
Customers Who Bought This Also Bought