Vivacon® 2 Centrifugal Concentrators, Sartorius
Supplier: Sartorius
Certificates
About this item
Vivacon® 2 centrifugal concentrators have been designed for DNA concentration, buffer exchange applications and primer removal.
- Optimal performance with very dilute samples for example, forensic specimens
- Low binding materials
- High recoveries even from diluted samples
- Re-spin possibility
- Complete and highly reproducible sample recovery
- Volume graduation and the molecular weight cut-off printed on individual devices
For samples up to 2 ml with regenerated cellulose Hydrosart® membrane. They feature a horizontal membrane design that is preferred by some users for reasons of consistency with previously used procedures and reproducibility.
Specifications
- Catalog No:
- CA76408-826
- CA76408-822
- CA76408-796
- CA76408-818
- CA76408-792
- CA76408-790
- CA76408-812
- CA76408-808
- CA76408-804
- CA76408-800
- CA76408-836
- CA76408-834
- Filtration area:
- 0.95 cm²
- 0.95 cm²
- 0.95 cm²
- 0.95 cm²
- 0.95 cm²
- 0.95 cm²
- 0.95 cm²
- 0.95 cm²
- 0.95 cm²
- 0.95 cm²
- 0.95 cm²
- 0.95 cm²
- Dead stop volume:
- 55 µl
- 55 µl
- 55 µl
- 55 µl
- 55 µl
- 55 µl
- 55 µl
- 55 µl
- 55 µl
- 55 µl
- 55 µl
- 55 µl
- Hold-up volume:
- 10 µl
- 10 µl
- 10 µl
- 10 µl
- 10 µl
- 10 µl
- 10 µl
- 10 µl
- 10 µl
- 10 µl
- 10 µl
- 10 µl