PIG® Straight-Sided Open-Head UN Rated Poly Drum, New Pig
Catalog #: 10860-600
Supplier: New Pig
Specifications
- Description:Straight-sided open-head UN rated poly drum, no bung holes
- Cat. no.:10860-600
- Capacity:55 gal.
- Dimensions:23Ø×37"H
- Supplier No.:DRM1059
Specifications
About this item
Straight sidewall, UN rated open top drum has ribs for extra stength and allows broader use of drum grabs and other equipment.
- Straight sidewalls allow broader use of drum grabs, parrot beak lifters and other equipment that can't move tapered drums
- Ribs add strength and rigidity, UN Rated for storage and transport of solid hazardous materials
- High-density polyethylene won't rust or corrode
- Quick-release lever lock band can be wire sealed to deter unauthorized access
Compatible with a wide range of liquids and chemicals.
Customers Who Bought This Also Bought