About this item
The J.T.Baker® BAKERBOND® PROchievA™ protein A resin offers superior dynamic binding capacity with excellent alkaline stability linked to Avantor's proprietary ligand.
- Available in 25 ml, 100 ml, 500 ml, 1 L and 5 L sizes, please contact your Avantor representative for pricing on the larger sizes
The J.T.Baker® BAKERBOND® PROchievA™ protein A resin is designed for high-performance in the critical affinity chromatography step used in the purification of mAbs, Fc fusion proteins, and other similar IgG-like molecules.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® PROchievA™ protein A resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot NumberAvantor Focus Production Volume 3 Issue 1 Avantor J.T.BAKER® BAKERBOND® PROchievA™ Recombinant Protein A Affinity Chromatography Resin VWR Protein Isolation and Purification VWR Top Rated Guide
Product Literature
Avantor J.T.Baker BAKERBOND PROchievA column packing guidelines Technical Note Avantor J.T.Baker BAKERBOND PROchievA pre-packed column guideline Avantor J.T.Baker Binding affinity of PROchievA resin to IgG-Technical Note Avantor J.T.Baker Specific binding to Fc domain of BAKERBOND PROchievA resin-Technical Note
Application Notes
spec_JT7899-03 spec_JT7899-05