About this item
A cost-effective, highly efficient auto-fill liquid nitrogen (LN2) tank.
- Auto-fill convenience
- Extremely low LN2 consumption
- Remote alarm contacts
- Vapor storage temperature of –185 °C (–301 °F) or colder
- VWR® two-year limited parts and labor and five-year limited vacuum warranty
Convenient design with reduced opening neck, hanging racks, and high vacuum insulation provide extremely low vapor temperatures of –185 °C (–301 °F) or colder using only a few inches of LN₂.
CryoRetriever claw-style tool is specifically designed for retrieving boxes, vials, and frames from cryopreservation tanks. Exclusive sealed center shaft prevents hazardous spurting of LN₂ that can occur with a hollow shaft retrieval claw.
Typically requires less than one LN₂ supply cylinder per month. Easy-to-use controller features digital temperature display, audible and visual low level alarms, low supply and high tank temperature alarms and remote alarm contacts. Unit is mounted on 5 wheeled caster platform and includes a locking provision. Under factor setting, 90% of sample capacity is in vapor phase storage.
Professional installation and setup is available and highly recommended for this product. Add this item to your order to receive on-site initial start-up (which includes controller settings, and personnel training by the manufacturer).
Cryostorage unit only includes seven stainless steel hanging racks. Each rack has a capacity of 10 boxes (not included). Package systems include cryostorage unit, full inventory system (stainless steel hanging racks, 100-cell boxes, and box dividers), 1.8 m (6') stainless steel transfer hose, liquid level measuring stick, and a pair of cryo-gloves.
Specifications
- Catalog No:
- 76411-300
- 76411-296
- 76411-298
- Canister height:
- 73.7 cm (Usable Internal Height)
- 73.7 cm (Usable Internal Height)
- 73.7 cm (Usable Internal Height)
- Neck int.Ø:
- 216 mm (8½")
- 216 mm (8½")
- 216 mm (8½")
- Static holding time (days):
- 194 days (nominal)
- 194 days (nominal)
- 194 days (nominal)
- Vial capacity:
- 7000×2 ml vials
- 7000×2 ml vials
- 7000×2 ml vials
- Weight Full:
- 186.4 kg (410 lbs)
- 186.4 kg (410 lbs)
- 186.4 kg (410 lbs)