Free Standing Biohazard Shield, TrippNT
Catalog #: 76419-596
Supplier: TrippNT
Specifications
- Length:16"
- Width:40"
- Height:13.375"
- Cat. no.:76419-596
- Weight:11 lbs.
Specifications
About this item
TrippNT's free standing biohazard shield is made from high-quality acrylic and HDPE to help prevent cross-contamination at work stations.
- Ships flat with easy snap-in assembly
- Works as biohazard shield or bench divider
- Clear acrylic construction
- Can be cleaned easily with wipes or mild detergents
The Biohazard shield can be easily disinfected with isopropyl alcohol or mild detergents, preventing cross-contamination between work stations.
Customers Who Bought This Also Bought