Digoxigenin-dUTP, Alkali Stable, Biotium
Catalog #: 75832-836
Supplier: Biotium
CAS Number:
Specifications
- Description:Digoxigenin-dUTP, alkali stable
- Size:25 nmol
- Storage conditions:–20°
- Cat. no.:75832-836
- Supplier No.:40078
Specifications
About this item
Digoxigenin-dUTP can be enzymatically incorporated into DNA via nick translation, random priming, or 3'-end terminal labeling to synthesize labeled DNA probes for in-situ hybridization, microarray or blotting techniques. The digoxigenin (DIG) labeled probe could be detected using a labeled anti-digoxigenin antibody.
Customers Who Bought This Also Bought