Specifications
- Description:Geno™ II 40 (US) Type 2 water system
- Environmentally Preferable:Sustainable Packaging,Sustainable Materials,Low Manufacturing Impact,Energy Efficient,Waste Reducing,Water Efficient
- Flow rate:0.67 L/min, 40 L/h
- Model:Geno™ 40
- Inorganics @ 25°C:1 - 15 MΩ-cm
- W×D×H:482.6×508×279.4 mm
- Bacteria:< 99 rejection
- Average daily needs:900 L/day
- Feed water:Tap water
- Resistivity at +25 °C:1 - 15 MΩ-cm
- Weight:25 kg
- Cat. no.:76533-358
Specifications
About this item
Provides laboratory grade, Type 2 water in two convenient production rates of 20 or 40 liters per hour.
- Type 2 (DI) Water
- RO Sanitization Process
- Simple operation with easy-change consumables
- Mount on bench, wall or on top of a reservoir
- Intuitive, icon-based, interactive user interface with touch screen
- Internal leak detector and boost pump included
- Choice of storage reservoir size - 30 L, 60 L, or 100 L
- Up to 25 or 45 L/hr production flow rates
Geno takes RO water to the next level of purity by providing laboratory grade, Type 2 water in two convenient production rates of 20 or 40 liters per hour. Select the reservoir size to suit your requirements. Geno combines all required technologies to produce Type 2 water in one box. The Endure DI packs are simple to change and long lasting. The integral boost pump simplifies installation and avoids unexpected costs. Rejection rate for particles: >99%. Feedwater pressure: 1.4 - 87 psi.
Additional: Geno is suitable for DI - Type 2 water usage of 10 - 600 liters per day. DI Water is suitable for buffer and media production, sample reagent diluents, general chemistry, protein electrophoresis, histology, cytology, and spectrophotometry.