Specifications
- Description:Chloride test 0 - 3000 mg/l
- Range:0 · 500 · 1000 · 1500 · 2000 · ≥3000 mg/l
- No. of Strips:100
Specifications
About this item
A ready-to-use kit that fits into your lab coat pocket.
- Rapid and convenient
- Easy to handle
Ideal to control your disinfection. Semi-quantitative results can be obtained between 10 and 120 seconds.
The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.